Assay Details
| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081060.1 | NP_001074529.1 | 11 - 12 | 1750 | 58 | |
| GenBank mRNA | AF139194.1 | - | 1 - 2 | 101 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3
Genomic Map