Assay Details
| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081060.1 | NP_001074529.1 | 13 - 14 | 2055 | 77 | |
| GenBank mRNA | AK033564.1 | - | 1 - 2 | 157 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3
Genomic Map