Assay Details
| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081060.1 | NP_001074529.1 | 2 - 3 | 501 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a3| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081060.1 | NP_001074529.1 | 2 - 3 | 501 | 67 | |
Set Membership: |
|