Assay Details
| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081060.1 | NP_001074529.1 | 7 - 8 | 1338 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a3| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081060.1 | NP_001074529.1 | 7 - 8 | 1338 | 97 | |
Set Membership: |
|