Assay Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027324.5 | NP_081600.1 | 3 - 4 | 436 | 69 | |
| GenBank mRNA | AF325260.1 | - | 3 - 4 | 414 | 69 | |
| AK012650.1 | - | 3 - 4 | 435 | 69 | ||
| AK151110.1 | - | 3 - 4 | 382 | 69 | ||
| BC005743.1 | - | 3 - 4 | 393 | 3599636 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sfxn1
Genomic Map