Assay Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027324.5 | NP_081600.1 | 5 - 6 | 616 | 63 | |
| GenBank mRNA | AF325260.1 | - | 5 - 6 | 594 | 63 | |
| AK012650.1 | - | 5 - 6 | 615 | 63 | ||
| AK151110.1 | - | 5 - 6 | 562 | 63 | ||
| BC005743.1 | - | 5 - 6 | 573 | 3599636 | 63 | |
More Information
Set Membership: |
|

Sfxn1
Genomic Map