Assay Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027324.5 | NP_081600.1 | 6 - 7 | 696 | 76 | |
| GenBank mRNA | AF325260.1 | - | 6 - 7 | 674 | 76 | |
| AK012650.1 | - | 6 - 7 | 695 | 76 | ||
| AK076205.1 | - | 1 - 2 | 65 | 76 | ||
| AK151110.1 | - | 6 - 7 | 642 | 76 | ||
| AK186603.1 | - | 1 - 2 | 66 | 76 | ||
| AK205405.1 | - | 1 - 2 | 66 | 76 | ||
| BC005743.1 | - | 6 - 7 | 653 | 3599636 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sfxn1
Genomic Map