Assay Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027324.5 | NP_081600.1 | 7 - 8 | 824 | 78 | |
| GenBank mRNA | AF325260.1 | - | 7 - 8 | 802 | 78 | |
| AK012650.1 | - | 7 - 8 | 823 | 78 | ||
| AK076205.1 | - | 2 - 3 | 193 | 78 | ||
| AK151110.1 | - | 7 - 8 | 770 | 78 | ||
| BC005743.1 | - | 7 - 8 | 781 | 3599636 | 78 | |
More Information
Set Membership: |
|

Sfxn1
Genomic Map