Assay Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027324.5 | NP_081600.1 | 8 - 9 | 875 | 92 | |
| GenBank mRNA | AF325260.1 | - | 8 - 9 | 853 | 92 | |
| AK012650.1 | - | 8 - 9 | 874 | 92 | ||
| AK076205.1 | - | 3 - 4 | 244 | 92 | ||
| AK151110.1 | - | 8 - 9 | 821 | 92 | ||
| BC005743.1 | - | 8 - 9 | 832 | 3599636 | 92 | |
More Information
Set Membership: |
|

Sfxn1
Genomic Map