Assay Details
| Gene Symbol: Porcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036746.1 | - | 4 - 5 | 548 | 55 | |
| AB036747.1 | - | 4 - 5 | 548 | 55 | ||
| AB036748.1 | - | 4 - 5 | 548 | 55 | ||
| AB036749.1 | - | 4 - 5 | 548 | 55 | ||
| BC027292.1 | - | 4 - 5 | 429 | 55 | ||
| BC032284.1 | - | 4 - 5 | 518 | 5362485 | 55 | |
| BQ556448.1 | - | 3 - 4 | 365 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Porcn
Genomic Map