Assay Details
| Gene Symbol: Ftsj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290430.1 | NP_001277359.1 | 11 - 12 | 1218 | 119 | |
| NM_133991.2 | NP_598752.2 | 11 - 12 | 1224 | 119 | ||
| GenBank mRNA | AK036313.1 | - | 11 - 12 | 1182 | 119 | |
| BC011144.1 | - | 10 - 11 | 1080 | 4194843 | 119 | |
| BC034880.1 | - | 11 - 12 | 1088 | 3468633 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ftsj1
Genomic Map