Assay Details
| Gene Symbol: Ftsj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290430.1 | NP_001277359.1 | 7 - 8 | 738 | 82 | |
| NM_133991.2 | NP_598752.2 | 7 - 8 | 738 | 82 | ||
| GenBank mRNA | AK036313.1 | - | 7 - 8 | 696 | 82 | |
| BC011144.1 | - | 6 - 7 | 642 | 4194843 | 82 | |
| BC034880.1 | - | 7 - 8 | 608 | 3468633 | 82 | |
More Information
Set Membership: |
|

Ftsj1
Genomic Map