Assay Details
| Gene Symbol: Lect2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010702.2 | NP_034832.2 | 3 - 4 | 463 | 144 | |
| GenBank mRNA | AB009687.1 | - | 3 - 4 | 425 | 144 | |
| AF035161.1 | - | 3 - 4 | 354 | 144 | ||
| AI173435.1 | - | 1 - 2 | 144 | |||
| AK131971.1 | - | 3 - 4 | 374 | 144 | ||
| BC027753.1 | - | 3 - 4 | 5049578 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074762 | AK131971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074762 | BC027753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074762 | AI173435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074762 | NM_010702.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074762 | AB009687.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134389 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166118 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074762 | AK131971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074762 | BC027753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074762 | AI173435.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074762 | NM_010702.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074762 | AB009687.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249909 | AK131971.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249909 | BC027753.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249909 | AI173435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249909 | NM_010702.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249909 | AB009687.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29061939 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166118 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07885770 | - | - |
| Whole Mouse Genome G4122A | A_51_P249909 | AK131971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249909 | BC027753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249909 | AI173435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249909 | NM_010702.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249909 | AB009687.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134914 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166118 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863287 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863307 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29061967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10572783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10621129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10776581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11102551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10863291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10863314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11102555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11102547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10956801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10936771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10936770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10863276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11102554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29061910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409596 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449492_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409592 | - | - |
| Mouse Genome 430 2.0 Array | 1449492_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1449492_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16841_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16841_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300013209.mm.1 | NM_010702.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013209.mm.1 | NM_010702.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013209.mm.1 | TC1300002083.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449492_PM_a_at | - | - |
| Mouse Expression Set 430 | 1449492_a_at | - | - |
| Murine Genome U74v2 Set | 102938_PM_at | - | - |
| Murine Genome U74v2 Set | 102938_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449492_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449492_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449492_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449492_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 50279 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000050279 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223634 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223634 | - | - |
| mouse-6 v1.1 (Illumina) | scl43907.5.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 50279 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43907.5.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 50279 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223634 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223634 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl43907.5.1_1-S | - | - |

Lect2
Genomic Map