Assay Details
| Gene Symbol: Shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040459.1 | NP_001035549.1 | 2 - 3 | 273 | 69 | |
| GenBank mRNA | BC151022.1 | - | 2 - 3 | 564 | 9087935 | 69 |
| DQ435686.1 | - | 2 - 3 | 273 | 69 | ||
| DQ435687.1 | - | 2 - 3 | 273 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shroom4
Genomic Map