Assay Details
| Gene Symbol: Shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040459.1 | NP_001035549.1 | 3 - 4 | 404 | 88 | |
| GenBank mRNA | AK052531.1 | - | 2 - 3 | 447 | 88 | |
| AK173129.1 | - | 2 - 3 | 426 | 88 | ||
| BC151022.1 | - | 3 - 4 | 695 | 9087935 | 88 | |
| DQ435686.1 | - | 3 - 4 | 404 | 88 | ||
| DQ435687.1 | - | 3 - 4 | 404 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shroom4
Genomic Map