Assay Details
| Gene Symbol: Morc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177719.4 | NP_808387.2 | 1 - 2 | 480 | 73 | |
| GenBank mRNA | AK077016.1 | - | 1 - 2 | 481 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077016.1, NM_177719.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110689 | BC120897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110689 | BC120896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110689 | AK077016.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110689 | NM_177719.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511521 | AK077016.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511521 | NM_177719.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511521 | AK077016.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511521 | NM_177719.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118344 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146285 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110689 | BC120897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110689 | BC120896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110689 | AK077016.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110689 | NM_177719.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511521 | AK077016.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511521 | NM_177719.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511521 | AK077016.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511521 | NM_177719.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148511 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146285 | - | - |
| Whole Mouse Genome G4122A | A_51_P511521 | AK077016.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511521 | NM_177719.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511521 | AK077016.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511521 | NM_177719.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102652 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146285 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13368565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13318548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224835 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13318550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13318543 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13368579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13236759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13423305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13357858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13360822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13368579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13318549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13358367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13236764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13368570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13358361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13475539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449988 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439222_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449979 | - | - |
| Mouse Genome 430 2.0 Array | 1439222_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439222_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240069_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240069_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700017258.mm.1 | NM_177719.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017258.mm.1 | NM_177719.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017258.mm.1 | TC1700001865.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017257.mm.1 | NM_177719.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017257.mm.1 | NM_177719.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017257.mm.1 | TC1700001865.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017260.mm.1 | NM_177719.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017260.mm.1 | NM_177719.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017260.mm.1 | TC1700001865.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017261.mm.1 | NM_177719.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017261.mm.1 | NM_177719.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017261.mm.1 | TC1700001865.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439222_PM_at | - | - |
| Mouse Expression Set 430 | 1439222_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439222_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439222_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 699697 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3290739 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3290739 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236692 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236692 | - | - |
| mouse-6 v1.1 (Illumina) | scl50050.3.1_186-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290739 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50050.3.1_186-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290739 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2872287 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2872287 | - | - |

Morc2b
Genomic Map