Assay Details
| Gene Symbol: Nxf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289735.1 | NP_001276664.1 | 14 - 15 | 1470 | 99 | |
| NM_001289736.1 | NP_001276665.1 | 14 - 15 | 1412 | 99 | ||
| NM_031259.2 | NP_112549.1 | 14 - 15 | 1470 | 99 | ||
| GenBank mRNA | AB178900.1 | - | 14 - 15 | 1470 | 99 | |
| AF285575.1 | - | 14 - 15 | 1470 | 99 | ||
| AY017476.1 | - | 14 - 15 | 1412 | 99 | ||
| BC137928.1 | - | 14 - 15 | 1469 | 8860947 | 99 | |
| BC137929.1 | - | 14 - 15 | 1469 | 8860948 | 99 | |
| BC145056.1 | - | 14 - 15 | 1469 | 9053604 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf2
Genomic Map