Assay Details
| Gene Symbol: Armcx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202500.3 | NP_001189429.2 | 3 - 4 | 289 | 72 | |
| GenBank mRNA | CJ167759.1 | - | 3 - 4 | 289 | 72 | |
| CJ177091.1 | - | 3 - 4 | 289 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001202500.3 transcripts.
Set Membership: |
|

Armcx4
Genomic Map