Assay Details
| Gene Symbol: Armcx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202500.3 | NP_001189429.2 | 4 - 5 | 374 | 93 | |
| GenBank mRNA | CJ167759.1 | - | 4 - 5 | 374 | 93 | |
| CJ177091.1 | - | 4 - 5 | 374 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001202500.3 transcripts.
Set Membership: |
|

Armcx4
Genomic Map