Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175214.3 | NP_780423.2 | 14 - 15 | 3360 | 114 | |
| GenBank mRNA | AK032912.1 | - | 7 - 8 | 1215 | 114 | |
| BC116646.1 | - | 12 - 13 | 2199 | 40086396 | 114 | |
More Information
Set Membership: |
|

Kif27
Genomic Map