Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175214.3 | NP_780423.2 | 4 - 5 | 1603 | 83 | |
| GenBank mRNA | BC116646.1 | - | 2 - 3 | 442 | 40086396 | 83 |
More Information
Set Membership: |
|

Kif27
Genomic Map