Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175214.3 | NP_780423.2 | 5 - 6 | 1809 | 111 | |
| GenBank mRNA | BC116646.1 | - | 3 - 4 | 648 | 40086396 | 111 |
More Information
Set Membership: |
|

Kif27
Genomic Map