| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300013344.mm.1 |
NM_175214.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013344.mm.1 |
NM_175214.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013344.mm.1 |
TC1300002097.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013343.mm.1 |
NM_175214.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013343.mm.1 |
NM_175214.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013343.mm.1 |
TC1300002097.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013342.mm.1 |
NM_175214.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013342.mm.1 |
NM_175214.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013342.mm.1 |
TC1300002097.mm.1 |
MTA TC |