Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175214.3 | NP_780423.2 | 8 - 9 | 2242 | 99 | |
| GenBank mRNA | AK032912.1 | - | 1 - 2 | 97 | 99 | |
| BC116646.1 | - | 6 - 7 | 1081 | 40086396 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif27
Genomic Map