Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175214.3 | NP_780423.2 | 9 - 10 | 2441 | 125 | |
| GenBank mRNA | AK032912.1 | - | 2 - 3 | 296 | 125 | |
| BC116646.1 | - | 7 - 8 | 1280 | 40086396 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif27
Genomic Map