Assay Details
| Gene Symbol: Dync1li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146229.2 | NP_666341.1 | 5 - 6 | 796 | 116 | |
| GenBank mRNA | AK159702.1 | - | 838 | 116 | ||
| AK189933.1 | - | 1 - 2 | 155 | 116 | ||
| AK191001.1 | - | 1 - 2 | 155 | 116 | ||
| AK191112.1 | - | 1 - 2 | 157 | 116 | ||
| AK191959.1 | - | 1 - 2 | 155 | 116 | ||
| AK193282.1 | - | 1 - 2 | 155 | 116 | ||
| AK194365.1 | - | 1 - 2 | 155 | 116 | ||
| AK195148.1 | - | 1 - 2 | 155 | 116 | ||
| AK195418.1 | - | 1 - 2 | 155 | 116 | ||
| AK196420.1 | - | 1 - 2 | 156 | 116 | ||
| AK196819.1 | - | 1 - 2 | 156 | 116 | ||
| AK197477.1 | - | 1 - 2 | 155 | 116 | ||
| AK197747.1 | - | 1 - 2 | 155 | 116 | ||
| AK198065.1 | - | 1 - 2 | 155 | 116 | ||
| AK204826.1 | - | 1 - 2 | 155 | 116 | ||
| AK213717.1 | - | 1 - 2 | 155 | 116 | ||
| AK218794.1 | - | 1 - 2 | 155 | 116 | ||
| BC023347.1 | - | 5 - 6 | 796 | 5040044 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dync1li1
Genomic Map