Assay Details
| Gene Symbol: Nr4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB221628.1 | - | 4 - 5 | 1461 | 116 | |
| AF191211.1 | - | 4 - 5 | 1461 | 116 | ||
| BC068150.1 | - | 6 - 7 | 2048 | 116 | ||
| BC128307.1 | - | 4 - 5 | 1456 | 116 | ||
| BC128308.1 | - | 4 - 5 | 1457 | 116 | ||
More Information
Set Membership: |
|

Nr4a3
Genomic Map