Assay Details
| Gene Symbol: Ctnnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018761.3 | NP_061231.3 | 13 - 14 | 1778 | 59 | |
| GenBank mRNA | AF006071.1 | - | 13 - 14 | 1765 | 59 | |
| BC050070.1 | - | 13 - 14 | 1755 | 30012833 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnal1
Genomic Map