Assay Details
| Gene Symbol: 4930520P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036596.1 | 2 - 3 | 251 | 94 | ||
| GenBank mRNA | AK015856.1 | - | 2 - 3 | 252 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 3772 | Gm3772 | -107.5kb | Sense | Chr. 13 - 70021959 - 70125917 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 1700100L14 gene | 1700100L14Rik | 328.5kb | Sense | Chr. 13 - 70573241 - 70576104 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930520P13Rik
Genomic Map