Assay Details
Gene Symbol: Ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007635.4 | NP_031661.3 | 5 - 6 | 827 | 86 | |
GenBank mRNA | AF005885.1 | - | 5 - 6 | 805 | 86 | |
AK041941.1 | - | 5 - 6 | 1145 | 86 | ||
AK168891.1 | - | 4 - 5 | 670 | 86 | ||
AK170651.1 | - | 5 - 6 | 811 | 86 | ||
AK198595.1 | - | 2 - 3 | 224 | 86 | ||
BC088980.1 | - | 5 - 6 | 633 | 5686564 | 86 | |
U95826.1 | - | 5 - 6 | 725 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
