Assay Details
| Gene Symbol: Mageh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023788.3 | NP_076277.1 | 1 | 1280 | 85 | |
| GenBank mRNA | AK008349.1 | - | 1 | 914 | 85 | |
| AK145866.1 | - | 1 | 1242 | 85 | ||
| BC051935.1 | - | 1 | 1069 | 5714800 | 85 | |
| BC058380.1 | - | 1 | 1059 | 6820349 | 85 | |
| BC060080.1 | - | 1 | 1069 | 5714800 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751752 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751750 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751751 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751748 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372039 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115942 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32751743 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115942 | - | - |
| Whole Mouse Genome G4122A | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | BC058380.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | BC060080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | AK000896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | AK008349.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497692 | AK145866.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120533 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14856418 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14856405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14856408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751748 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751750 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751751 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32751752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32751737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14561341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14856405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14681891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14856418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14700593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14809009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32751787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14561340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607396 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422498_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607395 | - | - |
| Mouse Genome 430 2.0 Array | 1422498_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422498_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75625_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75625_PM_at | - | - |
| Mouse Expression Set 430 | 1422498_PM_at | - | - |
| Mouse Expression Set 430 | 1422498_at | - | - |
| Murine Genome U74v2 Set | 160917_PM_r_at | - | - |
| Murine Genome U74v2 Set | 160917_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422498_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422498_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422498_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422498_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 324103 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6400102 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6400102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2767468 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234322 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2767468 | - | - |
| mouse-6 v1.1 (Illumina) | scl53701.1.378_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53701.1.378_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2996941 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2996941 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53701.1.378_12-S | - | - |

Mageh1
Genomic Map