Assay Details
| Gene Symbol: Rragb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004154.2 | NP_001004154.1 | 3 - 4 | 749 | 153 | |
| GenBank mRNA | BC069180.1 | - | 3 - 4 | 707 | 6810543 | 153 |
| BC070407.1 | - | 3 - 4 | 726 | 6808979 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rragb
Genomic Map