Assay Details
| Gene Symbol: Cxcl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009141.3 | NP_033167.2 | 2 - 3 | 395 | 76 | |
| GenBank mRNA | BC024392.1 | - | 2 - 3 | 307 | 4976854 | 76 |
| BQ829929.1 | - | 2 - 3 | 313 | 76 | ||
| U27267.1 | - | 2 - 3 | 315 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cxcl5
Genomic Map