Assay Details
| Gene Symbol: Rassf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028478.2 | NP_082754.2 | 5 - 6 | 523 | 59 | |
| GenBank mRNA | BC039934.1 | - | 5 - 6 | 534 | 59 | |
More Information
Set Membership: |
|

Rassf6
Genomic Map