Assay Details
| Gene Symbol: Areg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009704.3 | NP_033834.1 | 1 - 2 | 260 | 70 | |
| GenBank mRNA | AK018590.1 | - | 1 - 2 | 276 | 70 | |
| AK020273.1 | - | 1 - 2 | 276 | 70 | ||
| BC009138.1 | - | 1 - 2 | 260 | 3597695 | 70 | |
| CT010320.1 | - | 1 - 2 | 61 | 70 | ||
| D12648.1 | - | 1 - 2 | 87 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Areg
Genomic Map