Assay Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008918.1 | - | 5 - 6 | 851 | 82 | |
| AK030021.1 | - | 14 - 15 | 2141 | 82 | ||
| AK156076.1 | - | 14 - 15 | 2217 | 82 | ||
| AK168634.1 | - | 14 - 15 | 2174 | 82 | ||
| BC038877.1 | - | 2 - 3 | 287 | 82 | ||
| BC125391.1 | - | 14 - 15 | 2197 | 40129906 | 82 | |
| BC125393.1 | - | 14 - 15 | 2194 | 40129908 | 82 | |
| BC144795.1 | - | 14 - 15 | 2192 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Secisbp2
Genomic Map