Assay Details
| Gene Symbol: Auh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016709.2 | NP_057918.2 | 1 - 2 | 222 | 137 | |
| GenBank mRNA | AK003219.1 | - | 1 - 2 | 201 | 137 | |
| AK003929.1 | - | 1 - 2 | 202 | 137 | ||
| AK019978.1 | - | 1 - 2 | 199 | 137 | ||
| AK134886.1 | - | 1 - 2 | 194 | 137 | ||
| AK147855.1 | - | 1 - 2 | 226 | 137 | ||
| AK169661.1 | - | 1 - 2 | 200 | 137 | ||
| BC026525.1 | - | 1 - 2 | 160 | 137 | ||
| BC049597.1 | - | 1 - 2 | 206 | 6593786 | 137 | |
| BY741014.1 | - | 1 - 2 | 223 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Auh
Genomic Map