Assay Details
| Gene Symbol: Padi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011060.4 | NP_035190.3 | 10 - 11 | 1201 | 95 | |
| GenBank mRNA | AK220522.1 | - | 9 - 10 | 1443 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Padi3
Genomic Map