Assay Details
| Gene Symbol: Padi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011060.4 | NP_035190.3 | 2 - 3 | 312 | 134 | |
| GenBank mRNA | AK220522.1 | - | 1 - 2 | 554 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Padi3
Genomic Map