Assay Details
| Gene Symbol: Taf7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028958.4 | NP_083234.2 | 4 - 5 | 572 | 86 | |
| GenBank mRNA | AF285574.1 | - | 4 - 5 | 376 | 86 | |
| AK017109.1 | - | 4 - 5 | 573 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf7l
Genomic Map