Assay Details
| Gene Symbol: Was | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009515.2 | NP_033541.1 | 6 - 7 | 627 | 112 | |
| GenBank mRNA | U29673.1 | - | 6 - 7 | 588 | 112 | |
| U42471.1 | - | 6 - 7 | 588 | 112 | ||
| U54788.1 | - | 6 - 7 | 588 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Was
Genomic Map