Assay Details
| Gene Symbol: Epha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010139.3 | NP_034269.2 | 2 - 3 | 282 | 56 | |
| GenBank mRNA | AK137704.1 | - | 2 - 3 | 264 | 56 | |
| AK144202.1 | - | 2 - 3 | 266 | 56 | ||
| BC140960.1 | - | 2 - 3 | 238 | 9055572 | 56 | |
| BY024029.1 | - | 2 - 3 | 263 | 56 | ||
| CJ088701.1 | - | 2 - 3 | 286 | 56 | ||
| U07634.1 | - | 2 - 3 | 246 | 56 | ||
| X78339.1 | - | 2 - 3 | 244 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha2
Genomic Map