Assay Details
| Gene Symbol: Eif4ebp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201256.4 | NP_957708.1 | 1 - 2 | 243 | 62 | |
| GenBank mRNA | AY226182.1 | - | 1 - 2 | 121 | 62 | |
| BC038258.1 | - | 1 - 2 | 150 | 62 | ||
| BC061242.1 | - | 1 - 2 | 243 | 6743910 | 62 | |
| BC111107.1 | - | 1 - 2 | 183 | 40055518 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | AK129284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | AY226182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | BC038258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | NM_201256.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | BC061242.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | AK129284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | AY226182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | BC038258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | NM_201256.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P278497 | BC061242.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921320 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118698 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | AK129284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | AY226182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | BC038258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | NM_201256.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | BC061242.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | AK129284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | AY226182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | BC038258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | NM_201256.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P278497 | BC061242.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625116 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118698 | - | - |
| Whole Mouse Genome G4122A | A_51_P155604 | AK129284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155604 | AY226182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155604 | BC038258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155604 | NM_201256.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155604 | BC061242.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | AK129284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | AY226182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | BC038258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | NM_201256.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | BC061242.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | AK129284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | AY226182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | BC038258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | NM_201256.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278497 | BC061242.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133917 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118698 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13779076 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13886654 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13779078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13779077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13609613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13886656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13907170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625106 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10454947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454945 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454944 | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108112_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108112_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800002565.mm.1 | NM_201256.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002565.mm.1 | NM_201256.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002565.mm.1 | TC1800000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800002564.mm.1 | NM_201256.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002564.mm.1 | NM_201256.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002564.mm.1 | TC1800000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800002568.mm.1 | NM_201256.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002568.mm.1 | NM_201256.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002568.mm.1 | TC1800000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800002562.mm.1 | NM_201256.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002562.mm.1 | NM_201256.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002562.mm.1 | TC1800000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800002563.mm.1 | NM_201256.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002563.mm.1 | NM_201256.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002563.mm.1 | TC1800000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800002566.mm.1 | NM_201256.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002566.mm.1 | NM_201256.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002566.mm.1 | TC1800000332.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448008_at | - | - |
| Mouse Expression Set 430 | 1448008_PM_at | - | - |
| Mouse Expression Set 430 | 1419867_PM_a_at | - | - |
| Mouse Expression Set 430 | 1419867_a_at | - | - |
| Mouse Expression Set 430 | 1436597_PM_at | - | - |
| Mouse Expression Set 430 | 1453023_PM_at | - | - |
| Mouse Expression Set 430 | 1453023_at | - | - |
| Mouse Expression Set 430 | 1455759_a_at | - | - |
| Mouse Expression Set 430 | 1436597_at | - | - |
| Mouse Expression Set 430 | 1455759_PM_a_at | - | - |
| Murine Genome U74v2 Set | 103774_PM_at | - | - |
| Murine Genome U74v2 Set | 103774_at | - | - |
| Murine Genome U74v2 Set | 129318_at | - | - |
| Murine Genome U74v2 Set | 129318_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419867_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448008_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448008_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419867_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1230725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1230725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001230725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2701143 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2701143 | - | - |
| mouse-6 v1.1 (Illumina) | scl0108112.2_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1230725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0108112.2_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2895312 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2895312 | - | - |

Eif4ebp3
Genomic Map