Assay Details
| Gene Symbol: Vegfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025250.3 | NP_001020421.2 | 6 - 7 | 1511 | 65 | |
| NM_001110266.1 | NP_001103736.1 | 3 - 4 | 821 | 65 | ||
| NM_001287056.1 | NP_001273985.1 | 6 - 7 | 1511 | 65 | ||
| GenBank mRNA | AW913188.1 | - | 1 - 2 | 69 | 65 | |
| BQ554097.1 | - | 3 - 4 | 466 | 65 | ||
| S37052.1 | - | 6 - 7 | 502 | 65 | ||
More Information
Set Membership: |
|

Vegfa
Genomic Map