Assay Details
| Gene Symbol: Fam63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133858.4 | NP_598619.2 | 7 - 8 | 1974 | 75 | |
| NM_199475.1 | NP_955769.1 | 7 - 8 | 1974 | 75 | ||
| GenBank mRNA | AB075603.1 | - | 6 - 7 | 985 | 75 | |
| AK036838.1 | - | 7 - 8 | 1966 | 75 | ||
| AK169673.1 | - | 7 - 8 | 1974 | 75 | ||
| AK215644.1 | - | 2 - 3 | 75 | |||
| BC039762.1 | - | 6 - 7 | 792 | 75 | ||
| BC051048.1 | - | 6 - 7 | 857 | 75 | ||
| BC057901.1 | - | 5 - 6 | 503 | 75 | ||
| BC093521.1 | - | 3 - 4 | 543 | 4007022 | 75 | |
| BC132301.1 | - | 6 - 7 | 1219 | 40130578 | 75 | |
| BC144749.1 | - | 6 - 7 | 1219 | 9053297 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam63a
Genomic Map