Assay Details
| Gene Symbol: Fam63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133858.4 | NP_598619.2 | 5 - 6 | 1730 | 79 | |
| NM_199475.1 | NP_955769.1 | 5 - 6 | 1730 | 79 | ||
| GenBank mRNA | AB075603.1 | - | 4 - 5 | 741 | 79 | |
| AK036838.1 | - | 5 - 6 | 1722 | 79 | ||
| AK087859.1 | - | 5 - 6 | 1543 | 79 | ||
| AK169673.1 | - | 5 - 6 | 1730 | 79 | ||
| BC039762.1 | - | 4 - 5 | 548 | 79 | ||
| BC051048.1 | - | 4 - 5 | 613 | 79 | ||
| BC057901.1 | - | 3 - 4 | 259 | 79 | ||
| BC093521.1 | - | 1 - 2 | 299 | 4007022 | 79 | |
| BC094388.1 | - | 5 - 6 | 1547 | 5708548 | 79 | |
| BC132301.1 | - | 4 - 5 | 975 | 40130578 | 79 | |
| BC144749.1 | - | 4 - 5 | 975 | 9053297 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam63a
Genomic Map