Assay Details
| Gene Symbol: Fam63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133858.4 | NP_598619.2 | 8 - 9 | 2172 | 57 | |
| GenBank mRNA | AK036838.1 | - | 8 - 9 | 2164 | 57 | |
| BC008112.1 | - | 1 - 2 | 113 | 57 | ||
| BC057901.1 | - | 6 - 7 | 701 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam63a
Genomic Map