Assay Details
| Gene Symbol: Ccdc30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270435.1 | NP_001257364.1 | 11 - 12 | 1842 | 110 | |
| NM_001270446.1 | NP_001257375.1 | 6 - 7 | 1098 | 110 | ||
| NM_028506.1 | NP_082782.1 | 7 - 8 | 1105 | 110 | ||
| NM_029286.2 | NP_083562.1 | 11 - 12 | 1857 | 110 | ||
| GenBank mRNA | AK049010.1 | - | 6 - 7 | 998 | 110 | |
| AK078362.1 | - | 11 - 12 | 1784 | 110 | ||
| AK163185.1 | - | 11 - 12 | 1783 | 110 | ||
| AK224940.1 | - | 7 - 8 | 1034 | 110 | ||
| AK224961.1 | - | 6 - 7 | 1097 | 110 | ||
| BC114436.1 | - | 11 - 12 | 1784 | 40059415 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270446.1, NM_028506.1 transcripts.
Set Membership: |
|

Ccdc30
Genomic Map