Assay Details
| Gene Symbol: A530058N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028423.1 | 3 - 4 | 393 | 99 | ||
| GenBank mRNA | AK043120.1 | - | 3 - 4 | 243 | 95 | |
| AK045761.1 | - | 3 - 4 | 249 | 99 | ||
| AK045885.1 | - | 3 - 4 | 198 | 99 | ||
| AK046235.1 | - | 3 - 4 | 223 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
gap junction protein, delta 2 |
Gjd2 |
Gja9, connexin36, cx36 |
-6.6kb | Anti-sense | Chr. 2 - 114009601 - 114013619 on NCBI Build GRCm38 |
| Downstream |
actin, alpha, cardiac muscle 1 |
Actc1 | Actc-1 | 24.0kb | Anti-sense | Chr. 2 - 114047289 - 114052811 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A530058N18Rik
Genomic Map