Assay Details
| Gene Symbol: Slc5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133254.3 | NP_573517.1 | 11 - 12 | 1441 | 90 | |
| GenBank mRNA | AK128900.1 | - | 11 - 12 | 1455 | 90 | |
| AY033886.1 | - | 11 - 12 | 1451 | 90 | ||
| BC022226.1 | - | 11 - 12 | 1441 | 4235707 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a2
Genomic Map